Novus Biologicals
Manufacturer Code:NB10074365
Catalog # NB10074365
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | CC-4 |
Immunogen | Amino acid residues 123-229 of human BAP31. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: 6C6-AG tumor-associated antigen; B-cell receptor-associated protein 31; Bap316C6-AG BAP31DXS1357ECDM B-cell receptor-associated protein 31 BCR-associated protein 31 BCR-associated protein Bap316C6-AG tumor-associated antigen p28 p28 Bap316C6-Ag Protein CDM; BCR-associated protein 31; BCR-associated protein Bap31; p28; p28 Bap31; Protein CDM
Gene Aliases: 6C6-AG; BAP31; BCAP31; CDM; DDCH; DXS1357E
UniProt ID: (Human) P51572
Entrez Gene ID: (Human) 10134
Molecular Function: membrane traffic protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.