Novus Biologicals
Manufacturer Code:NBP272368PCP
Catalog # NB272368PCP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7A11 |
Immunogen | Full length recombinant protein of human BCAP/PIK3AP1 (NP_689552) produced in HEK293T cells. |
Conjugate | PerCP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 1810044J04Rik B cell adaptor protein BCAPB-cell adapter for phosphoinositide 3-kinase B-cell phosphoinositide 3-kinase adapter protein 1 FLJ35564 phosphoinositide 3-kinase adapter protein 1 phosphoinositide-3-kinase adaptor protein 1 RP11-34E5.3; B cell adaptor protein; B-cell adapter for phosphoinositide 3-kinase; B-cell phosphoinositide 3-kinase adapter protein 1; Phosphoinositide 3-kinase adapter protein 1
Gene Aliases: BCAP; PIK3AP1
UniProt ID: (Human) Q6ZUJ8
Entrez Gene ID: (Human) 118788
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.