Novus Biologicals
Manufacturer Code:NBP297928H
Catalog # NB049555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human BBX fragment. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ag recognized by Treg cells 1; Bobby sox homolog; Bobby sox homolog bobby sox homolog (Drosophila) HBP2HSPC339 HMG box transcription factor BBX HMG box-containing protein 2 MDS001 x 001 protein; HMG box transcription factor BBX; HMG box-containing protein 2; x 001 protein
Gene Aliases: ARTC1; BBX; HBP2; HSPC339; MDS001
UniProt ID: (Human) Q8WY36
Entrez Gene ID: (Human) 56987
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.