Invitrogen
Manufacturer Code:PA554373
Catalog # PIPA554373
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human BBS9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bardet-Biedl syndrome 9 protein; Parathyroid hormone-responsive B1 gene protein; Protein PTHB1; Protein PTHB1 Bardet-Biedl syndrome 9 protein Parathyroid hormone-responsive B1 gene protein BBS9 PTHB1; PTH-responsive osteosarcoma B1 protein
Gene Aliases: B1; BBS9; C18; D1; PTHB1
UniProt ID: (Human) Q3SYG4
Entrez Gene ID: (Human) 27241
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.