Invitrogen
Manufacturer Code:PA5114245
Catalog # PIPA5114245
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal region of Human BBS9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B1 Bardet-Biedl syndrome 9 Bardet-Biedl syndrome 9 (human) Bardet-Biedl syndrome 9 protein Bardet-Biedl syndrome 9 protein homolog BBS9 C18 D1 E130103I17Rik EST 3159894 Parathyroid hormone-responsive B1 gene protein Parathyroid hormone-responsive B1 gene protein homolog Protein PTHB1 PTHB1 PTH-responsive osteosarcoma B1 protein; Bardet-Biedl syndrome 9 protein; Parathyroid hormone-responsive B1 gene protein; Protein PTHB1; PTH-responsive osteosarcoma B1 protein
Gene Aliases: B1; BBS9; C18; D1; PTHB1
UniProt ID: (Human) Q3SYG4
Entrez Gene ID: (Human) 27241
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.