Novus Biologicals
Manufacturer Code:NBP188927
Catalog # NBP188927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:QEDTQELGWEETVDAAISHLLKTCLSKSSKEQALNLNSQLNIPKDTSQLKKHITLLCDRLSKGGRLCLSTDAAAPQTMVMPGGCTTIPESDLEERSVEQDSTELFTNHRHLTAETPRPEVSPLQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B1 Bardet-Biedl syndrome 9 Bardet-Biedl syndrome 9 protein C18 D1 Parathyroid hormone-responsive B1 gene protein protein PTHB1 PTHB1MGC118917 PTH-responsive osteosarcoma B1 protein; Bardet-Biedl syndrome 9 protein; Parathyroid hormone-responsive B1 gene protein; Protein PTHB1; PTH-responsive osteosarcoma B1 protein
Gene Aliases: B1; BBS9; C18; D1; PTHB1
UniProt ID: (Human) Q3SYG4
Entrez Gene ID: (Human) 27241
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.