Proteintech
Manufacturer Code:144601AP150UL
Catalog # 50-172-9467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BBS9 Fusion Protein Ag5665 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B1 Bardet Biedl syndrome 9 BBS9 C18 D1 Protein PTHB1 PTHB1; Bardet-Biedl syndrome 9 protein; Bardet-Biedl syndrome 9 protein homolog; EST 3159894; Parathyroid hormone-responsive B1 gene protein; Parathyroid hormone-responsive B1 gene protein homolog; Protein PTHB1; PTH-responsive osteosarcoma B1 protein
Gene Aliases: B1; BBS9; C18; D1; E130103I17Rik; PTHB1
UniProt ID: (Human) Q3SYG4, (Mouse) Q8BKD7
Entrez Gene ID: (Human) 27241, (Mouse) 319845
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.