Invitrogen
Manufacturer Code:PA522055
Catalog # PIPA522055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 263 and 540 of BBS7 (Uniprot IDNo.Q8IWZ6) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bardet-Biedl syndrome 7 protein; Bardet-Biedl syndrome 7 protein homolog; BBS2-like 1; BBS2-like protein 1; BBS2L1 BBS2-like 1 BBS2-like protein 1 Bardet-Biedl syndrome 7 protein
Gene Aliases: 8430406N16Rik; BBS2L1; BBS7
UniProt ID: (Human) Q8IWZ6, (Mouse) Q8K2G4
Entrez Gene ID: (Human) 55212, (Mouse) 71492
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.