Proteintech
Manufacturer Code:189611AP150UL
Catalog # 50-173-2290
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BBS7 Fusion Protein Ag4601 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bardet Biedl syndrome 7 BBS2 like protein 1 BBS2L1 BBS7; Bardet-Biedl syndrome 7 protein; Bardet-Biedl syndrome 7 protein homolog; BBS2-like 1; BBS2-like protein 1
Gene Aliases: 8430406N16Rik; BBS2L1; BBS7
UniProt ID: (Human) Q8IWZ6, (Mouse) Q8K2G4
Entrez Gene ID: (Human) 55212, (Mouse) 71492
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.