Abnova
Manufacturer Code:H00055212K
Catalog # 89-201-968
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human BBS7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human BBS7 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Bardet-Biedl syndrome 7 protein; BBS2-like 1; BBS2-like protein 1; BBS2L1 FLJ10715
Gene Aliases: BBS2L1; BBS7
UniProt ID: (Human) Q8IWZ6
Entrez Gene ID: (Human) 55212
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.