Proteintech
Manufacturer Code:130781AP150UL
Catalog # 50-172-8472
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BBS6 Fusion Protein Ag3785 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bardet-Biedl syndrome 6 protein; BBS6 HMCS KMS McKusick Kaufman syndrome MKKS MKS; McKusick-Kaufman syndrome protein; McKusick-Kaufman syndrome protein; MKKS protein; McKusick-Kaufman/Bardet-Biedl syndromes putative chaperonin; Protein Bbs6 homolog
Gene Aliases: 1300013E18Rik; AI463362; AI957237; BBS6; HMCS; KMS; MKKS; MKS
UniProt ID: (Human) Q9NPJ1, (Mouse) Q9JI70
Entrez Gene ID: (Human) 8195, (Mouse) 59030, (Rat) 311456
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.