Proteintech
Manufacturer Code:661361IG150UL
Catalog # 50-173-6572
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G8E12 |
Immunogen | BBS5 Fusion Protein Ag6844 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Bardet Biedl syndrome 5 BBS5; Bardet-Biedl syndrome 5 protein; Bardet-Biedl syndrome 5 protein homolog; novel DUF1448 protein
Gene Aliases: 1700049I01Rik; 2700023J09Rik; BBS5
UniProt ID: (Human) Q8N3I7, (Mouse) Q9CZQ9
Entrez Gene ID: (Human) 129880, (Mouse) 72569
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.