Invitrogen
Manufacturer Code:PA5113979
Catalog # PIPA5113979
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human BBS5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700049I01Rik 2700023J09Rik Bardet-Biedl syndrome 5 Bardet-Biedl syndrome 5 (human) Bardet-Biedl syndrome 5 homolog Bardet-Biedl syndrome 5 protein Bardet-Biedl syndrome 5 protein homolog Bbs5 novel DUF1448 protein; Bardet-Biedl syndrome 5 protein
Gene Aliases: BBS5
UniProt ID: (Human) Q8N3I7
Entrez Gene ID: (Human) 129880
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.