Novus Biologicals
Manufacturer Code:NBP270241UV
Catalog # NBP270241UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D5 |
Immunogen | Recombinant protein fragment corresponding to amino acids 266-519 of human BBS4 (NP_149017) produced in E.coli. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Bardet-Biedl syndrome 4 Bardet-Biedl syndrome 4 protein; Bardet-Biedl syndrome 4 protein
Gene Aliases: BBS4
UniProt ID: (Human) Q96RK4
Entrez Gene ID: (Human) 585
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.