Novus Biologicals
Manufacturer Code:NBP232308
Catalog # NBP232308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1292CT845.130.218 |
Immunogen | This BBS4 antibody is generated from a mice immunized with a recombinant protein between 1-240 amino acids from human BBS4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Bardet-Biedl syndrome 4 Bardet-Biedl syndrome 4 protein; Bardet-Biedl syndrome 4 protein
Gene Aliases: BBS4
UniProt ID: (Human) Q96RK4
Entrez Gene ID: (Human) 585
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.