Invitrogen
Manufacturer Code:PA5104328
Catalog # PIPA5104328
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BBS4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW537059 AW742241 Bardet-Biedl syndrome 4 Bardet-Biedl syndrome 4 (human) Bardet-Biedl syndrome 4 homolog Bardet-Biedl syndrome 4 protein Bardet-Biedl syndrome 4 protein homolog Bbs4 D9Ertd464e; Bardet-Biedl syndrome 4 homolog; Bardet-Biedl syndrome 4 protein; Bardet-Biedl syndrome 4 protein homolog
Gene Aliases: AW537059; AW742241; BBS4; D9Ertd464e
UniProt ID: (Human) Q96RK4, (Mouse) Q8C1Z7
Entrez Gene ID: (Human) 585, (Mouse) 102774, (Rat) 300754
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.