Invitrogen
Manufacturer Code:PA545024
Catalog # PIPA545024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human BBS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bardet-Biedl syndrome 2 BBS MGC20703; Bardet-Biedl syndrome 2 homolog; Bardet-Biedl syndrome 2 protein; Bardet-Biedl syndrome 2 protein homolog
Gene Aliases: 2410125H22Rik; AI447581; BBS; BBS2; fb80a05; RP74; wu:fb80a05
UniProt ID: (Human) Q9BXC9, (Rat) Q99MH9, (Mouse) Q9CWF6, (Zebrafish) Q98SP7
Entrez Gene ID: (Dog) 478121, (Human) 583, (Horse) 100051424, (Rabbit) 100343044, (Bovine) 533611, (Guinea pig) 100728338, (Rat) 113948, (Mouse) 67378, (Zebrafish) 259187
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.