Novus Biologicals
Manufacturer Code:NBP169082
Catalog # NBP169082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to BBS2 (Bardet-Biedl syndrome 2) The peptide sequence was selected from the N terminal of BBS2. Peptide sequence GSDLFWTVTGDNVNSLALCDFDGDGKKELLVGSEDFDIRVFKEDEIVAEM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bardet-Biedl syndrome 2 Bardet-Biedl syndrome 2 protein BBS MGC20703; Bardet-Biedl syndrome 2 protein
Gene Aliases: BBS; BBS2; RP74
UniProt ID: (Human) Q9BXC9
Entrez Gene ID: (Human) 583
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.