Proteintech
Manufacturer Code:124212AP150UL
Catalog # 50-172-7970
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BBS10 Fusion Protein Ag3073 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bardet Biedl syndrome 10 BBS10 C12orf58; Bardet-Biedl syndrome 10 protein
Gene Aliases: BBS10; C12orf58; si:dkey-30c15.16
UniProt ID: (Human) Q8TAM1
Entrez Gene ID: (Human) 79738, (Zebrafish) 100034535
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.