Proteintech
Manufacturer Code:211181AP150UL
Catalog # 50-173-3265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BBS1 Fusion Protein Ag15460 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bardet Biedl syndrome 1 BBS1 BBS2 like protein 2 BBS2L2; Bardet-Biedl syndrome 1 homolog; Bardet-Biedl syndrome 1 protein; Bardet-Biedl syndrome 1 protein homolog; BBS2-like protein 2
Gene Aliases: AI451249; BBS1; BBS2L2; D19Ertd609e
UniProt ID: (Human) Q8NFJ9, (Mouse) Q3V3N7
Entrez Gene ID: (Human) 582, (Mouse) 52028
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.