Novus Biologicals
Manufacturer Code:NBP29198820UL
Catalog # NB025870
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein of Human BAZ2A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bromodomain adjacent to zinc finger domain protein 2A; bromodomain adjacent to zinc finger domain protein 2A bromodomain adjacent to zinc finger domain 2A DKFZp781B109 FLJ45876 hWALp3 KIAA0314FLJ13768 Tip5 TIP5FLJ13780 Transcription termination factor I-interacting protein 5 TTF-I interacting peptide 5 TTF-I-interacting protein 5 WALp3; bromodomain adjacent to zinc finger domain, 2A; hWALp3; Transcription termination factor I-interacting protein 5; TTF-I interacting peptide 5; TTF-I-interacting protein 5
Gene Aliases: BAZ2A; KIAA0314; TIP5; WALp3
UniProt ID: (Human) Q9UIF9
Entrez Gene ID: (Human) 11176
Molecular Function:
DNA binding protein
acetyltransferase
chromatin/chromatin-binding protein
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.