R&D Systems
Manufacturer Code:MAB7437
Catalog # MAB7437
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 841702 |
Immunogen | <EM>E. coli</EM>-derived recombinant human BATF3 Met1-Arg127 Accession # Q9NR55 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Ms Monoclonal Antibody
Protein Aliases: 21 kDa small nuclear factor isolated from T-cells; 21-kD small nuclear factor isolated from T cells; B-ATF-3; basic leucine zipper transcription factor ATF-like 3 basic leucine zipper transcriptional factor ATF-like 3 B-ATF-3 FLJ36352 FLJ3753521 kDa small nuclear factor isolated from T-cells JDP1 Jun dimerization protein 1 Jun dimerization protein p21SNFT JUNDM1 SNFT SNFT21-kD small nuclear factor isolated from T cells; basic leucine zipper transcription factor, ATF-like 3; Basic leucine zipper transcriptional factor ATF-like 3; Jun dimerization protein 1; Jun dimerization protein p21SNFT
Gene Aliases: BATF3; JDP1; JUNDM1; SNFT
UniProt ID: (Human) Q9NR55
Entrez Gene ID: (Human) 55509
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.