Invitrogen
Manufacturer Code:PA578526
Catalog # PIPA578526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full length human BATF3 Recombit protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 21 kDa small nuclear factor isolated from T-cells; 21-kD small nuclear factor isolated from T cells; B-ATF-3; basic leucine zipper transcription factor, ATF-like 3; Basic leucine zipper transcriptional factor ATF-like 3; Jun dimerization protein 1; Jun dimerization protein p21SNFT; JUNDM1 JDP1 SNFT basic leucine zipper transcription factor ATF-like 3
Gene Aliases: BATF3; JDP1; JUNDM1; SNFT
UniProt ID: (Human) Q9NR55
Entrez Gene ID: (Human) 55509
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.