Abnova
Manufacturer Code:H00055509K
Catalog # 89-201-958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human BATF3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human BATF3 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 21 kDa small nuclear factor isolated from T-cells; 21-kD small nuclear factor isolated from T cells; B-ATF-3; basic leucine zipper transcription factor, ATF-like 3; Basic leucine zipper transcriptional factor ATF-like 3; JDP1 JUNDM1 SNFT; Jun dimerization protein 1; Jun dimerization protein p21SNFT
Gene Aliases: BATF3; JDP1; JUNDM1; SNFT
UniProt ID: (Human) Q9NR55
Entrez Gene ID: (Human) 55509
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.