R&D Systems
Manufacturer Code:FAB8054T100UG
Catalog # FAB8054T100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 687706 |
Immunogen | <EM>E. coli</EM>-derived recombinant human BATF Met1-Pro125 Accession # Q16520 |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: activating transcription factor B; B-ATF; B-cell-activating transcription factor; basic leucine zipper transcription factor ATF-like BATF1basic leucine zipper transcriptional factor ATF-like B-ATFSFA2 B-cell-activating transcription factor SFA2 SFA-2activating transcription factor B SF-HT-activated gene 2 protein; basic leucine zipper transcription factor, ATF-like; Basic leucine zipper transcriptional factor ATF-like; SF-HT-activated gene 2 protein; SFA-2
Gene Aliases: B-ATF; BATF; BATF1; SFA-2; SFA2
UniProt ID: (Human) Q16520
Entrez Gene ID: (Human) 10538
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.