Invitrogen
Manufacturer Code:PA540595
Catalog # PIPA540595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human BATF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activating transcription factor B; B-ATF; B-cell activating transcription factor; B-cell-activating transcription factor; Basic leucine zipper transcription factor ATF-like B-ATF BATF1 SFA-2 SFA2; basic leucine zipper transcription factor, ATF-like; basic leucine zipper transcription factor-like; Basic leucine zipper transcriptional factor ATF-like; SF-HT-activated gene 2 protein; SFA-2
Gene Aliases: B-ATF; BATF; BATF1; SFA-2; SFA2; si:ch211-153j24.4
UniProt ID: (Human) Q16520, (Bovine) E1BD44, (Rat) D4A7E1, (Mouse) O35284, (Zebrafish) F1QW76
Entrez Gene ID: (Rabbit) 100338089, (Horse) 100057690, (Human) 10538, (Bovine) 617628, (Dog) 611951, (Rat) 299206, (Mouse) 53314, (Guinea pig) 100714129, (Zebrafish) 564589
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.