Proteintech
Manufacturer Code:135071AP150UL
Catalog # 50-172-8794
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BATF Fusion Protein Ag4418 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activating transcription factor B; B ATF BATF BATF1 SF HT activated gene 2 protein SFA 2 SFA2; B-ATF; B-cell activating transcription factor; B-cell-activating transcription factor; basic leucine zipper transcription factor, ATF-like; basic leucine zipper transcription factor-like; Basic leucine zipper transcriptional factor ATF-like; SF-HT-activated gene 2 protein; SFA-2
Gene Aliases: B-ATF; BATF; BATF1; SFA-2; SFA2
UniProt ID: (Human) Q16520, (Rat) D4A7E1, (Mouse) O35284
Entrez Gene ID: (Human) 10538, (Rat) 299206, (Mouse) 53314
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.