Invitrogen
Manufacturer Code:PA545048
Catalog # PIPA545048
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human BASP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 22 kDa neuronal tissue-enriched acidic protein; Brain abundant membrane attached signal protein 1 CAP-23 CAP23 MGC8555 NAP-22 NAP22; brain abundant, membrane attached signal protein 1; Brain acid soluble protein 1; brain acid-soluble protein 1; brain acidic membrane protein; Neuronal axonal membrane protein NAP-22; neuronal tissue-enriched acidic protein
Gene Aliases: BASP1; CAP-23; CAP23; NAP-22; NAP22
UniProt ID: (Human) P80723, (Rat) Q05175
Entrez Gene ID: (Human) 10409, (Rabbit) 100351504, (Rat) 64160
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.