Invitrogen
Manufacturer Code:PA5111311
Catalog # PIPA5111311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human BASP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 22 kDa neuronal tissue-enriched acidic protein; 22 kDa neuronal tissue-enriched acidic protein 2610024P12Rik Basp1 Brain acid soluble protein 1 CAP23 CAP-23 Ckap3 MGC8555 NAP22 NAP-22 Neuronal axonal membrane protein NAP-22; brain abundant, membrane attached signal protein 1; Brain acid soluble protein 1; brain acid-soluble protein 1; Neuronal axonal membrane protein NAP-22; neuronal tissue-enriched acidic protein
Gene Aliases: BASP1; CAP-23; CAP23; NAP-22; NAP22
UniProt ID: (Human) P80723
Entrez Gene ID: (Human) 10409
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.