Novus Biologicals
Manufacturer Code:NBP146356
Catalog # NBP146356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from n-terminal region of mouse Brain acid soluble protein 1 (BASP1) conjugated to an immunogenic carrier protein was used as the antigen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 22 kDa neuronal tissue-enriched acidic protein; brain abundant membrane attached signal protein 122 kDa neuronal tissue-enriched acidic protein brain acid-soluble protein 1 CAP-23 CAP23brain acid soluble protein 1 NAP-22 NAP22MGC8555 Neuronal axonal membrane protein NAP-22 neuronal tissue-enriched acidic protein; brain abundant, membrane attached signal protein 1; Brain acid soluble protein 1; brain acid-soluble protein 1; Neuronal axonal membrane protein NAP-22; neuronal tissue-enriched acidic protein
Gene Aliases: BASP1; CAP-23; CAP23; NAP-22; NAP22
UniProt ID: (Human) P80723
Entrez Gene ID: (Human) 10409
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.