Novus Biologicals
Manufacturer Code:NBP299048AF405
Catalog # NB057554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human BASP1. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 22 kDa neuronal tissue-enriched acidic protein; brain abundant membrane attached signal protein 122 kDa neuronal tissue-enriched acidic protein brain acid-soluble protein 1 CAP-23 CAP23brain acid soluble protein 1 NAP-22 NAP22MGC8555 Neuronal axonal membrane protein NAP-22 neuronal tissue-enriched acidic protein; brain abundant, membrane attached signal protein 1; Brain acid soluble protein 1; brain acid-soluble protein 1; Neuronal axonal membrane protein NAP-22; neuronal tissue-enriched acidic protein
Gene Aliases: BASP1; CAP-23; CAP23; NAP-22; NAP22
UniProt ID: (Human) P80723
Entrez Gene ID: (Human) 10409
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.