Invitrogen
Manufacturer Code:PA540622
Catalog # PIPA540622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human BARX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BarH-like homeobox 1; BARX homeobox 1; Homeobox protein BarH-like 1
Gene Aliases: BARX1
UniProt ID: (Human) Q96GH8, (Mouse) Q9ER42
Entrez Gene ID: (Pig) 100512774, (Dog) 610486, (Rabbit) 100356518, (Human) 56033, (Rat) 364680, (Mouse) 12022, (Guinea pig) 100721978
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.