Invitrogen
Manufacturer Code:PA575304
Catalog # PIPA575304
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the N-terminual of Human BARD1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BARD-1; BRCA1-associated RING domain gene 1; BRCA1-associated RING domain protein 1; BRCA1-associated RING domain protein 1 BARD-1 BARD 1; RING-type E3 ubiquitin transferase BARD1
Gene Aliases: BARD1; ENSMUSG00000060893; ENSMUSG00000073653
UniProt ID: (Human) Q99728, (Mouse) A2VCQ1, (Rat) Q9QZH2
Entrez Gene ID: (Human) 580, (Mouse) 12021, (Rat) 64557
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.