Invitrogen
Manufacturer Code:PA565149
Catalog # PIPA565149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human BARD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BARD-1; BARD-1 BRCA1 associated RING domain 1 isoform alfa BRCA1 associated RING domain 1 isoform beta BRCA1 associated RING domain 1 isoform delta BRCA1 associated RING domain 1 isoform epsilon BRCA1 associated RING domain 1 isoform eta BRCA1 associated RING dom; BRCA1-associated RING domain gene 1; BRCA1-associated RING domain protein 1; RING-type E3 ubiquitin transferase BARD1
Gene Aliases: BARD1
UniProt ID: (Human) Q99728
Entrez Gene ID: (Human) 580
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.