Invitrogen
Manufacturer Code:PA578856
Catalog # PIPA578856
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human BAP1 recombit protein (Position: M1-E278). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brca1 associated protein 1; BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase); BRCA1-associated protein 1; Cerebral protein 6; cerebral protein-13; Ubiquitin C-terminal hydrolase X4; Ubiquitin carboxyl-terminal hydrolase BAP1; Ubiquitin carboxyl-terminal hydrolase BAP1 BRCA1-associated protein 1 Cerebral protein 6 BAP1 KIAA0272 hucep-6; UCH-X4
Gene Aliases: 2300006C11Rik; AA989761; AW553466; BAP1; HUCEP-13; hucep-6; KIAA0272; mKIAA0272; uch-x4; UCHL2
UniProt ID: (Human) Q92560, (Rat) D3ZHS6, (Mouse) Q99PU7
Entrez Gene ID: (Human) 8314, (Rat) 306257, (Mouse) 104416
Molecular Function:
cysteine protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.