Novus Biologicals
Manufacturer Code:NBP297934IR
Catalog # NB049614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human BANP fragment. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BEN domain containing 1 BEND1BEN domain-containing protein 1 BTG3 associated nuclear protein Btg3-associated nuclear protein DKFZp761H172 FLJ10177 FLJ20538 protein BANP SMAR1Scaffold/matrix-associated region-1-binding protein SMARBP1; BEN domain-containing protein 1; Btg3-associated nuclear protein; Protein BANP; Scaffold/matrix-associated region-1-binding protein
Gene Aliases: BANP; BEND1; SMAR1; SMARBP1
UniProt ID: (Human) Q8N9N5
Entrez Gene ID: (Human) 54971
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.