Abnova
Manufacturer Code:PAB19312
Catalog # 89-120-845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 14 amino acids near internal region of human BANP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody rasied against synthetic peptide of BANP.
For Research Use Only
Protein Aliases: BEN domain-containing protein 1; BEND1; Btg3-associated nuclear protein; DKFZp761H172; FLJ10177; FLJ20538; Protein BANP; Scaffold/matrix-associated region-1-binding protein; SMAR1; SMARBP1
Gene Aliases: BANP; BEND1; SMAR1; SMARBP1
UniProt ID: (Human) Q8N9N5
Entrez Gene ID: (Human) 54971
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.