Proteintech
Manufacturer Code:163931AP150UL
Catalog # 50-173-0916
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BANP Fusion Protein Ag9484 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BANP BEND1 Protein BANP SMAR1 SMARBP1; BEN domain-containing protein 1; Btg3-associated nuclear protein; Protein BANP; Scaffold/matrix-associated region-1-binding protein
Gene Aliases: AA408158; BANP; BEND1; SMAR1; SMARBP1
UniProt ID: (Human) Q8N9N5, (Mouse) Q8VBU8
Entrez Gene ID: (Human) 54971, (Rat) 292064, (Mouse) 53325
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.