Invitrogen
Manufacturer Code:PA558710
Catalog # PIPA558710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human BANF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAF BCRP1 D14S1460 NGPS barrier-to-autointegration factor breakpoint cluster region protein 1; Barrier-to-autointegration factor; Barrier-to-autointegration factor, N-terminally processed; Breakpoint cluster region protein 1
Gene Aliases: BAF; BANF1; BCRG1; BCRP1; D14S1460; NGPS
UniProt ID: (Human) O75531
Entrez Gene ID: (Human) 8815
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.