Invitrogen
Manufacturer Code:PA520328
Catalog # PIPA520328
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide from near the carboxy terminus of human BANF1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAF BCRP1 D14S1460 NGPS barrier-to-autointegration factor breakpoint cluster region protein 1; Barrier-to-autointegration factor; Barrier-to-autointegration factor, N-terminally processed; Breakpoint cluster region protein 1; LAP2-binding protein 1
Gene Aliases: BAF; BANF1; BCRG1; BCRP1; D14S1460; L2bp1; L2bp1/Baf; NGPS
UniProt ID: (Human) O75531, (Mouse) O54962
Entrez Gene ID: (Human) 8815, (Mouse) 23825
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.