Novus Biologicals
Manufacturer Code:NBP271778AF594
Catalog # NP271778A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5D5 |
Immunogen | Recombinant protein fragment corresponding to amino acids 21-152 of human BAMBI/NMA (NP_036474) produced in E.coli. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: BMP and activin membrane-bound inhibitor homolog; BMP and activin membrane-bound inhibitor homolog BMP and activin membrane-bound inhibitor homolog (Xenopus laevis) NMANon-metastatic gene A protein Putative transmembrane protein NMA; Non-metastatic gene A protein; Putative transmembrane protein NMA
Gene Aliases: BAMBI; NMA
UniProt ID: (Human) Q13145
Entrez Gene ID: (Human) 25805
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.