R&D Systems
Manufacturer Code:AF2387
Catalog # AF2387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse BAMBI/NMA Ile28-Thr143 Accession # Q9D0L6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP and activin membrane-bound inhibitor homolog; BMP and activin membrane-bound inhibitor homolog BMP and activin membrane-bound inhibitor homolog (Xenopus laevis) NMA NMANon-metastatic gene A protein Putative transmembrane protein NMA; Non-metastatic gene A protein; Putative transmembrane protein NMA
Gene Aliases: BAMBI; NMA
UniProt ID: (Human) Q13145
Entrez Gene ID: (Human) 25805
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.