Novus Biologicals
Manufacturer Code:NBP267460
Catalog # NBP267460
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SU32-07 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Apoptosis regulator BAK; Apoptosis regulator BAK BAK-LIKE BAKMGC3887 BCL2-antagonist/killer 1 Bcl2-L-7 BCL2L7bcl-2 homologous antagonist/killer BCL2-like 7 protein Bcl-2-like protein 7 CDN1 MGC117255 pro-apoptotic protein BAK; Bcl-2 homologous antagonist/killer; Bcl-2-like protein 7; BCL2-antagonist/killer 1; Bcl2-L-7; BCL2-like 7 protein; pro-apoptotic protein BAK
Gene Aliases: BAK; BAK-LIKE; BAK1; BCL2L7; CDN1
UniProt ID: (Human) Q16611
Entrez Gene ID: (Human) 578
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.