Invitrogen
Manufacturer Code:PA5106855
Catalog # PIPA5106855
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BAK1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis regulator BAK; Apoptosis regulator BAK Bak Bak1 BAK-LIKE BCL2 antagonist/killer 1 Bcl2 homologous antagonist/killer bcl-2 homologous antagonist/killer BCL2-antagonist/killer 1 BCL2L7 bcl2-L-7 BCL2-like 7 protein BCL2-like 7protein Bcl-2-like protein 7 Bcl-2-likeous antagonist/killer CDN1 MGC117255 MGC3887 N-Bak N-BAK1 pro-apoptotic protein BAK; Bcl-2 homologous antagonist/killer; Bcl-2-like protein 7; Bcl2 homologous antagonist/killer; BCL2-antagonist/killer 1; Bcl2-L-7; BCL2-like 7 protein; pro-apoptotic protein BAK
Gene Aliases: BAK; BAK-LIKE; BAK1; BCL2L7; CDN1; N-Bak; N-BAK1
UniProt ID: (Human) Q16611, (Mouse) O08734
Entrez Gene ID: (Human) 578, (Mouse) 12018, (Rat) 116502
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.