Novus Biologicals
Manufacturer Code:NB10056092SS
Catalog # NB10056092T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 16-36 (EPALPSASEEQVAQDTEEVFR) of human Bak was used as immunogen GenBank no. NP_001179.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis regulator BAK; Apoptosis regulator BAK BAK-LIKE BAKMGC3887 BCL2-antagonist/killer 1 Bcl2-L-7 BCL2L7bcl-2 homologous antagonist/killer BCL2-like 7 protein Bcl-2-like protein 7 CDN1 MGC117255 pro-apoptotic protein BAK; Bcl-2 homologous antagonist/killer; Bcl-2-like protein 7; BCL2-antagonist/killer 1; Bcl2-L-7; BCL2-like 7 protein; pro-apoptotic protein BAK
Gene Aliases: BAK; BAK-LIKE; BAK1; BCL2L7; CDN1
UniProt ID: (Human) Q16611
Entrez Gene ID: (Human) 578
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.