Abnova
Manufacturer Code:PAB20156
Catalog # 89-121-659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human BAIAP2L2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant BAIAP2L2.
For Research Use Only
Protein Aliases: BAI1-associated protein 2-like 2; BAI1-associated protein 2-like protein 2; Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 2; FLJ22582; Pinkbar; Planar intestinal- and kidney-specific BAR domain protein
Gene Aliases: BAIAP2L2; UNQ9336/PRO34007
UniProt ID: (Human) Q6UXY1
Entrez Gene ID: (Human) 80115
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.