Invitrogen
Manufacturer Code:PA522026
Catalog # PIPA522026
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 266 of BAIAP2L1 (Uniprot IDNo.Q9UHR4) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAI1-associated protein 2-like 1; BAI1-associated protein 2-like protein 1; Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 1; Insulin receptor tyrosine kinase substrate; IRTKS BAI1-associated protein 2-like protein 1 brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 1 insulin receptor tyrosine kinase substrate
Gene Aliases: BAIAP2L1; IRTKS
UniProt ID: (Human) Q9UHR4
Entrez Gene ID: (Human) 55971
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.