Invitrogen
Manufacturer Code:PA5114892
Catalog # PIPA5114892
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BAIAP2L1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1300006M19Rik AI585895 BAI1 associated protein 2 like 1 BAI1-associated protein 2-like 1 BAI1-associated protein 2-like protein 1 Baiap2l1 brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 1 Insulin receptor tyrosine kinase substrate Irtks RGD1308452; BAI1-associated protein 2-like 1; BAI1-associated protein 2-like protein 1; Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 1; Insulin receptor tyrosine kinase substrate
Gene Aliases: 1300006M19Rik; AI585895; BAIAP2L1; IRTKS; RGD1308452
UniProt ID: (Human) Q9UHR4, (Mouse) Q9DBJ3, (Rat) Q3KR97
Entrez Gene ID: (Human) 55971, (Mouse) 66898, (Rat) 304282
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.