Novus Biologicals
Manufacturer Code:NBP200810
Catalog # NBP200810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI 1D9 |
Immunogen | Full length human recombinant protein of human BAIAP2 (NP_006331) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BAI-associated protein 2; BAI1-associated protein 2; BAI1-associated protein 2IRS-58 BAI-associated protein 2 BAP2 brain-specific angiogenesis inhibitor 1-associated protein 2 Fas ligand-associated factor 3 FLAF3 Insulin receptor substrate p53 Insulin receptor substrate p53/p58 Insulin receptor substrate protein of 53 kDa IRSp53 IRSp53/58 Protein BAP2; Brain-specific angiogenesis inhibitor 1-associated protein 2; Fas ligand-associated factor 3; FLAF3; insulin receptor substrate of 53 kDa; Insulin receptor substrate p53/p58; Insulin receptor substrate protein of 53 kDa; IRS-58; IRSp53; IRSp53/58
Gene Aliases: BAIAP2; BAP2; FLAF3; IRSP53
UniProt ID: (Human) Q9UQB8
Entrez Gene ID: (Human) 10458
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.