R&D Systems
Manufacturer Code:FAB9338U100UG
Catalog # FAB9338U100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2528B |
Immunogen | Human embryonic kidney cell HEK293-derived human BAI2 Phe21-Asp911 Accession &num O60241-3 |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Adhesion G protein-coupled receptor B2; Brain-specific angiogenesis inhibitor 2; brain-specific angiogenesis inhibitor 2 Brain-specific angiongenesis inhibitor-2
Gene Aliases: ADGRB2; BAI2
UniProt ID: (Human) O60241
Entrez Gene ID: (Human) 576
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.